VWR® Basic ESD Mob Caps
113-8899
- Ø:530 mm
- Anti-static:+
- Garment application:General Purpose,Electronics/semi-conductor,Industrial,Food
- Colour:White
- Pk:1.000
- Weight:10 g/m²
These disposable pleated caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Single elastic
- Anti-static properties
Weight: 10 g/m²
: 100 pieces per bag, 10 bags per case.