Order Entry
Austria
ContactUsLinkComponent
 
1,1,1,3,3,3-Hexafluoro-2-propanol
Catalog # MOLEM24492514
Supplier:  Molekula
CAS Number:  
1,1,1,3,3,3-Hexafluoro-2-propanol
Catalog # MOLEM24492514
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:13,09598
Boiling Pt:59.1 °C (1013 hPa)
Melting Pt:–96 °C
Density:1.492 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:213-059-4
UN:1760
ADR:8,II
 :168.04 g/mol
 :GHS05
 :GHS07
CAS Number:920-66-1
Formula:C₃H₂F₆O
MDL Number:MFCD00011651