1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:920-66-1
:
Formula:C₃H₂F₆O
MDL Number:MFCD00011651
Merck Index:13,09598
Boiling Pt:59…60 °C
Melting Pt:–4 °C
Density:1.619
Flash Pt:>100 °C
UN:1760
ADR:8,II
:168.04 g/mol
EINECS:213-059-4
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together