

Oxalic acid, anhydrous for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:144-62-7
:
Formula:C₂H₂O₄
MDL Number:MFCD00002573
:90.04 g/mol
Merck Index:13,06980
Boiling Pt:365 °C (1013 hPa)
Melting Pt:99 - 100 °C
Density:1.9 g/cm³ (20 °C)
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:205-634-3
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:90 g/l (20 °C)
- Melting Point:189 °C (decomposition)
- Molar Mass:90.03 g/mol
- Bulk Density:750 kg/m3
- Vapor Pressure:21.5 hPa (50 °C)
- Density:1.9 g/cm3 (20 °C)
- pH:1 (100 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Frequently Bought Together