Glutaric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-94-1
:
Formula:C₅H₈O₄
MDL Number:MFCD00004410
:132.12 g/mol
Merck Index:13,04486
Boiling Pt:302 °C (1013 hPa)
Melting Pt:98 °C
Density:1.43 g/cm³ (15 °C)
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:203-817-2
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:640 g/l (20 °C)
- Melting Point:98 °C
- Molar Mass:132.11 g/mol
- Boiling Point:302 - 304 °C (1013 hPa) (slow decomposition)
- Vapor Pressure:0.022 hPa (18.5 °C)
- Density:1.43 g/cm3 (15 °C)
- Synonyms:Pentanedioic acid
- Hill Formula:C5H8O4
- Chemical Formula:HOOC(CH2)3COOH
Specification Test Results
Frequently Bought Together