Glutaric acid
Catalog # EHERC14034500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:132.12 g/mol
Merck Index:13,04486
Boiling Pt:302 °C (1013 hPa)
Melting Pt:98 °C
Density:1.43 g/cm³ (15 °C)
Storage Temperature:Ambient
EINECS:203-817-2
:GHS07
CAS Number:110-94-1
Formula:C₅H₈O₄
MDL Number:MFCD00004410