
Glutaric acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-94-1
:
Formula:C₅H₈O₄
MDL Number:MFCD00004410
:132.12 g/mol
Merck Index:13,04486
Boiling Pt:302 °C (1013 hPa)
Melting Pt:92…99 °C
Density:1,43 g/cm³ (15 °C)
EINECS:203-817-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:95 to 99 °C
- Titration with NaOH:98.5 to 101.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together