
Succinic acid 99.0-100.5% (by acidimetry), crystalline, EMPROVE® ESSENTIAL NF, JPE, ACS, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:110-15-6
:
Formula:HOOCCH₂CH₂COOH
MDL Number:MFCD00002789
:118.09 g/mol
Merck Index:13,08953
Boiling Pt:235 °C (1013 hPa)
Melting Pt:185 …190 °C
Density:1.56 g/cm³ (20 °C)
Flash Pt:206 °C
Storage Temperature:Ambient
EINECS:203-740-4
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Assay (acidimetric):99.0 − 100.5%
- Assay (HPLC):99.0 − 100.5%
- Identity:Conforms
- Appearance of solution (5%; water):Conforms
- In water insoluble matter:≤0.01%
- Melting range (lower value):≥185.0 °C
- Melting range (upper value):≤190.0 °C
- Melting point:185.0 − 190.0 °C
- Chloride (Cl):≤0.001%
- Phosphate (PO₄):≤0.001%
- Sulfate (SO₄):≤0.003%
- Heavy metals (as Pb):≤0.0020%
- Heavy metals (as Pb) (ACS):≤5 ppm
- Nitrogen compounds (as N):≤0.001%
- Fe (Iron):≤5 ppm
- As (Arsenic):≤0.0002%
- Substances reducing permanganate:Conforms
- Residual solvents (ICH Q3C):Excluded by production process
- Sulfated ash (600 °C):≤0.02%
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.00681.5000_TDS_EN ProductPage_100681
Product specification
msds_100681
SDS
Frequently Bought Together