Order Entry
Austria
ContactUsLinkComponent
L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Lieferant:  Avantor
Zertifikate
undefined
L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Lieferant:  Avantor
Zertifikate

Spezifikation Testergebnisse

  • GMP Manufactured Product:
    98.5 - 101.5%
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets E.P. Chemical Specifications:
    22.4 - 24.7 Degree
  • Meets J.P. Chemical Specifications:
    5.6 - 6.1
  • Meets U.S.P Requirements:
    ≤0.3%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.4%
  • Bulk Pharmaceutical Chemical:
    ≤0.05%
  • USP - Assay (C₅H₁₁NO₂S) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    22.4 - 24.7 Degree
  • USP - pH:
    5.6 - 6.1
  • USP - Loss on Drying at 105°C:
    ≤0.3 %
  • USP - Residue on Ignition:
    ≤0.4 %
  • USP - Chloride (Cl):
    ≤0.05 %
  • USP - Sulfate (SO₄):
    ≤0.03 %
  • USP - Iron (Fe):
    ≤0.003 %
  • USP - Related Compounds - Methionine sulfoxide:
    ≤0.1 %
  • USP - Related Compounds - N-Acetyl-D,L- methionine:
    ≤0.2 %
  • USP - Related Compounds - Any unspecified impurity:
    ≤0.1 %
  • USP - Related Compounds - Total unspecified impurities:
    ≤0.30 %
  • EP/BP - Assay (C₅H₁₁NO₂S) (dried basis):
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    5.5 - 6.5
  • EP/BP - Specific Rotation [Å]²º^D (+):
    22.5 - 24.0 Degree
  • EP/BP - Related Substances: Unspecified Impurities, each:
    ≤0.10 %
  • EP/BP - Related Substances: Total Impurities:
    ≤0.3 %
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.5 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JP - Assay (C₅H₁₁NO₂S) (dried basis):
    98.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation (+):
    21.0 - 25.0 Degree
  • JP - pH:
    5.2 - 6.2
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.021 %
  • JP - Sulfate (SO₄):
    ≤0.028 %
  • JP - Ammonium (NH₄):
    ≤0.02 %
  • JP - Heavy Metals (as Pb):
    ≤20 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤0.30 %
  • JP - Residue after Ignition:
    ≤0.10 %

Spezifikation Testergebnisse

Zugehörige Informationen

Frequently Bought Together