6-Aminocaproic acid, Sigma-Aldrich®
: Merck
:60-32-2
:
:C₆H₁₃NO₂
:MFCD00008238
:131.17 g/mol
:13,00431
:257 °C (1013 hPa)
:127 °C
:M-Clarity™ quality level = MQ200
:207 °C
:200-469-3
:GHS07
:Ambient
Frequently Bought Together
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Petroleum spirit, 60…95 °C, TECHNICAL
: VWR Chemicals