6-Aminocaproic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-32-2
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00008238
:131.17 g/mol
Merck Index:14,432
Boiling Pt:257 °C (1013 hPa)
Melting Pt:210…212 °C
Flash Pt:207 °C
EINECS:200-469-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together