Order Entry
Austria
ContactUsLinkComponent
6-Aminocaproic acid 99+%
6-Aminocaproic acid 99+%
Certificates
undefined
6-Aminocaproic acid 99+%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    204 to 209 °C (Dec)
  • Solubility:
    (13g/100ml H2O) Clear colorless solution
  • Titration with HClO4:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together