Sodium acetate, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt:881,4 °C (1013 hPa)
Melting Pt:324 °C
Density:1,528 g/cm³ (20 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤1200 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Frequently Bought Together