Potassium acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Frequently Bought Together