Sodium butyrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:156-54-7
:
Formula:C₄H₇NaO₂
MDL Number:MFCD00002816
:110.09 g/mol
Melting Pt:244 °C
Density:1.324 g/cm³ (30 °C)
:M-Clarity™ quality level = MQ200
EINECS:205-857-6
Frequently Bought Together