
Lauric acid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:143-07-7
:
Formula:C₁₂H₂₄O₂
MDL Number:MFCD00002736
:200.32 g/mol
Merck Index:13,05400
Boiling Pt:296 °C (1013 hPa)
Melting Pt:42…45 °C
Density:0.875 g/cm³ (20 °C)
Flash Pt:>150 °C
EINECS:205-582-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together