Order Entry
Austria
ContactUsLinkComponent
 
Triethyl citrate 99.0-101.0% (alkalimetric, calculated on anhydrous substance) Ph. Eur., JPE, NF, SAFC®
 
Triethyl citrate 99.0-101.0% (alkalimetric, calculated on anhydrous substance) Ph. Eur., JPE, NF, SAFC®
 

 

  • Assay (alkalimetric, calculated on anhydrous substance):
    99.0 - 101.0 %
  • Assay (GC, calc. on anhydrous substance):
    99.0 - 100.5 %
  • Identity (IR-spectrum):
    passes test
  • Identity (GC):
    passes test
  • Identity (wet chemistry):
    passes test
  • Identity (Refractive index):
    passes test
  • Appearance:
    clear solution, not more intense in color than matching fluid A (1:10) and reference solution BY₆
  • Acidity:
    passes test
  • Refractive index (n 20/D):
    1.440 - 1.446
  • Refractive index (n 25/D):
    1.439 - 1.441
  • Density (d 25/25):
    1.135 - 1.139
  • Density (d 20/20):
    1.135 - 1.145
  • Viscosity (20 °C):
    26 - 38 mPa∙s
  • Sulfate (SO₄):
    ≤ 150 ppm
  • Heavy metals (as Pb):
    ≤ 5 ppm
  • As (Arsenic):
    ≤ 2 ppm
  • Cyclohexane (HS-GC):
    ≤ 3880 ppm
  • Ethanol (HS-GC):
    ≤ 5000 ppm
  • Other residual solvents (ICH (Q3C)):
    excluded by manufacturing process
  • Related substances (GC) (largest single impurity):
    ≤ 0.2 %
  • Related substances (GC) (Sum of all impurities):
    ≤ 0.5 %
  • Triethyl aconitate (GC):
    ≤ 0.2 %
  • Related substances (GC):
    passes test
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Water (according to Karl Fischer):
    ≤ 0.25 %

 

Frequently Bought Together