Triethyl citrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-93-0
:
Formula:C₁₂H₂₀O₇
MDL Number:MFCD00009201
:276.29 g/mol
Boiling Pt:294 °C (1013 hPa)
Melting Pt:–55 °C
Density:1.135 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:150 °C
EINECS:201-070-7
Specification Test Results
- Solubility:65 g/l (25 °C)
- Melting Point:-55 °C
- Molar Mass:276.28 g/mol
- Boiling Point:294 °C (1013 hPa)
- Vapor Pressure:0.0025 hPa (25 °C)
- Flash Point:150 °C
- Density:1.135 g/cm3 (25 °C)
Specification Test Results
Frequently Bought Together