Order Entry
Austria
ContactUsLinkComponent
Triethyl citrate
Triethyl citrate
Catalog # EDQMT1986000
Supplier:  EDQM
CAS Number:  
Triethyl citrate
Catalog # EDQMT1986000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :276.29 g/mol
Boiling Pt:294 °C (1013 hPa)
Melting Pt:–55 °C
Density:1.135 g/cm³ (25 °C)
Flash Pt:150 °C
EINECS:201-070-7
CAS Number:77-93-0
Formula:C₁₂H₂₀O₇
MDL Number:MFCD00009201