Triethyl citrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-93-0
:
Formula:C₁₂H₂₀O₇
MDL Number:MFCD00009201
:276.29 g/mol
Boiling Pt:294 °C (760 mmHg)
Melting Pt:–46 °C
Density:1.136 g/cm³
Flash Pt:151 °C
Storage Temperature:Ambient
EINECS:201-070-7
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4430 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together