Dimethyl pimelate, Sigma-Aldrich®
Catalog # SIAL180068-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:250 °C (1013 hPa)
Melting Pt:–18.7 °C
Density:1.041 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:188.22 g/mol
Storage Temperature:Ambient
EINECS:217-057-4
CAS Number:1732-08-7
Formula:C₉H₁₆O₄
MDL Number:MFCD00008470