Dimethyl adipate for synthesis, Sigma-Aldrich®
: Merck
:627-93-0
:
:C₈H₁₄O₄
:MFCD00008469
:174.2 g/mol
:13,00163
:∼229 °C (1013 hPa)
:10.3 °C
:1.06 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:107 °C
:Ambient
:211-020-6
- Solubility:- 1 g/l (20 °C)
- Melting Point:8 - 11 °C
- Molar Mass:174.19 g/mol
- Boiling Point:215 - 225 °C (1013 hPa)
- Vapor Pressure:0.06 hPa (20 °C)
- Flash Point:107 °C
- Refractive Index:1.4285 (20 °C, 589 nm)
- Explosion Limit:0.81 - 8.1 %(V)
- Density:1.06 g/cm3 (20 °C)
- Ignition Point:360 °C
- Viscosity kinematic:2.5 mm2/s (25 °C)
- Synonyms:Dimethyl hexanedioate, Adipic acid dimethyl ester
- Hill Formula:C8H14O4
Frequently Bought Together