Diethyl succinate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-25-1
:
Formula:C₈H₁₄O₄
MDL Number:MFCD00009208
:174.2 g/mol
Merck Index:12,09038
Boiling Pt:217 °C (1013 hPa)
Melting Pt:–19 °C
Density:1.04 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:204-612-0
Frequently Bought Together