Order Entry
Austria
ContactUsLinkComponent
Diethyl succinate for synthesis, Sigma-Aldrich®
Diethyl succinate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Diethyl succinate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    2.00 g/l (25 °C)
  • Melting Point:
    -20 °C
  • Molar Mass:
    174.19 g/mol
  • Boiling Point:
    215 - 217 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (55 °C)
  • Flash Point:
    80 °C
  • Refractive Index:
    1.42 (20 °C, 589 nm)
  • Density:
    1.04 g/cm3 (20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together