Order Entry
Austria
ContactUsLinkComponent
Diethyl succinate 98%
Diethyl succinate 98%
Certificates
undefined
Diethyl succinate 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4180 to 1.4220 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together