Dimethyl sebacate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-79-6
:
Formula:C₁₂H₂₂O₄
MDL Number:MFCD00008472
:230.3 g/mol
Boiling Pt:287…289 °C
Melting Pt:25…28 °C
Density:0.988
Flash Pt:145 °C (293 °F)
Storage Temperature:Ambient
EINECS:203-431-4
Related Information
Frequently Bought Together