Order Entry
Austria
ContactUsLinkComponent
 
Ethyl octanoate for synthesis, Sigma-Aldrich®
 :  Merck
 
Ethyl octanoate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    0.070 g/l (25 °C)
  • Melting Point:
    -47 °C
  • Molar Mass:
    172.27 g/mol
  • Boiling Point:
    208 °C (1013 hPa)
  • Vapor Pressure:
    0.2 hPa (20 °C)
  • Flash Point:
    79 °C
  • Refractive Index:
    1.4166 (20 °C, 589 nm)
  • Explosion Limit:
    0.7 %(V)
  • Density:
    0.87 g/cm3 (20 °C)
  • Ignition Point:
    325 °C
  • Synonyms:
    Ethyl caprylate, Octanoic acid ethyl ester
  • Hill Formula:
    C10H20O2
  • Chemical Formula:
    CH3(CH2)6COOC2H5

 

Frequently Bought Together