Order Entry
Austria
ContactUsLinkComponent
Multi-parameter meters, handheld, Eutech CyberScan PCD 650
Multi-parameter meters, handheld, Eutech CyberScan PCD 650
Catalog # EUTEECPCDWP65044K
Supplier:  EUTECH
undefined
Multi-parameter meters, handheld, Eutech CyberScan PCD 650
Catalog # EUTEECPCDWP65044K
Supplier:  EUTECH

Specifications

  • Temperature range:
    −10,0...+110,0 °C
  • Description:
    Multi-parameter meters, handheld, Eutech CyberScan PCD 650
  • Model:
    PCD 650
  • ISE accuracy:
    0,5 % full scale (monovalent); 1 % full scale (divalent)
  • Temperature accuracy:
    ±0,5 °C
  • Temperature resolution:
    0,1 °C
  • DO range:
    0,00 - 90,00 mg/l
  • DO resolution:
    0,01 mg/l; 0,1 %
  • Compliance:
    IP67
  • Conductivity accuracy:
    ±1 % full scale
  • Conductivity calibration:
    4 points (1 per range) auto, 5 points (1 per range) manual
  • Conductivity cell constant:
    0,010 - 10,000 cm⁻¹
  • Conductivity range:
    0 - 500 mS/cm
  • Conductivity resolution:
    0,01 µS/cm; 0,1 µS/cm; 0,001 mS/cm; 0,01 mS/cm; 0,1 mS/cm
  • Connections:
    DC phono sockets, BNC, 8 pin connector, 6 pin connector
  • DO barometric pressure range:
    450 - 825 mmHg or 59,9 - 109,9 kPa
  • DO calibration:
    1 point
  • DO saturation:
    0 - 600,0 %
  • DO accuracy:
    ±0,2 mg/l
  • Data logging capacity:
    Up to 500 data sets
  • Display:
    With backlight
  • ISE calibration:
    Up to 8 points
  • ISE resolution:
    2 - 3 digits
  • ISE range:
    0,001 - 19900
  • Outputs:
    IrDA, RS232C (LED)
  • Power supply:
    4× 1,5 V AA alkaline batteries or 9 V DC adapter, 500 mA
  • Salinity accuracy:
    ±1 % full scale
  • Salinity range:
    0 - 80,00 ppt
  • Salinity resolution:
    0,01 ppm / 0,1 ppm / 0,001 ppt / 0,01 ppt
  • TDS accuracy:
    ±1% full scale
  • TDS range:
    0 - 500,0 ppt
  • TDS resolution:
    0,01 ppm / 0,1 ppm / 0,001 ppt / 0,01 ppt / 0,1 ppt
  • Temperature compensation:
    ATC/MTC (0...100 °C)
  • Weight:
    0,46 kg
  • W×D×H:
    95×57×183 mm
  • mV accuracy:
    ±0,2
  • mV range:
    ±2000,0
  • mV resolution:
    0,1
  • pH accuracy:
    ±0,002
  • pH calibration:
    1 (offset) to 6 points
  • pH range:
    –2,000...+20,000
  • pH resolution:
    0,1 / 0,01 / 0,001
  • Pk:
    1

Specifications

About this item

This portable meter enables users to measure pH, mV, conductivity, TDS, salinity, ions, dissolved oxygen and temperature values. It simultaneously measures and displays up to four parameters.

  • Accepts two-cell and four-cell conductivity probe to measure up to 500 m/s — meter measures pure water
  • Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
  • Electrode status report, calibration data and simultaneous display of up to 4 parameters
  • Rugged and IP67-rated waterproof for applications in harsh environments; meter floats on water
  • Recorded readings individually stamped with time and date in GLP compliant format
  • HI-LO limit alarm for easy quality control – Meter alerts when readings fall outside selected ranges
  • Wireless Information Technology: No wires, no cables — Transfer data from meter to computer with the press of a button

Ideal for environmental testing, water quality testing, industrial applications and aquaculture.

Delivery: Supplied with pH electrode ECFC7252203B, 4-cell conductivity electrode ECCONSEN9203J, dissolved oxygen probe ECDOHANDYNEW (all electrodes come with 3 m cables), DAS software, multi-probe holder, power adapter and carrying kit set incl. a plastic carrying case, 1× pH 4.01, pH 7.00 buffer solutions, storage solution and 1× deionised (rinse) water (60 ml each).