Orders LinkContactUsLinkComponent
Bio-N⁶-ddATP
Bio-N⁶-ddATP, 25 nmol, 25 Nmo
Catalog # ENZOENZ42809
Supplier:   Enzo Life Sciences
Bio-N⁶-ddATP
Catalog # ENZOENZ42809
Supplier:  Enzo Life Sciences
CAS Number:  
Bio-N⁶-ddATP
Bio-N⁶-ddATP
Bio-N⁶-ddATP, 25 nmol, 25 Nmo
CAS Number:  
Price
388,00 €
Log in for availability
1 * 25 Nmo
Product Details & Documents
Bio-N⁶-ddATP (Biotin-N⁶-(2’,3’-dideoxy)adenosine-5’ triphosphate) can be used for 3’-end labelling of single-stranded or double-stranded DNA. It is especially useful as a substrate for terminal transferase for the 3’-terminal addition of a single Bio-N⁶-ddAMP. It is also a substrate for Taq DNA Polymerase and other DNA polymerases.
Biotin-labelled oligonucleotides produced by this reaction are suitable as hybridisation probes. Biotin-labelled cDNA produced by this reaction can be used in nucleic acid array assays. Biotin-labelled DNA in hybridisation assays can be detected by a reporter molecule linked to streptavidin, avidin, or and anti-biotin antibody. Such a complex can be detected directly, by the excitation of a fluorophore conjugated to streptavidin, or indirectly by using an enzyme conjugate that can produce an insoluble coloured precipitate.
Specifications
Specifications
  • Pk:
    25 Nmo
  • Description:
    Bio-N⁶-ddATP, 25 nmol
Specifications