Ethyl laurate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-33-2
:
Formula:C₁₄H₂₈O₂
MDL Number:MFCD00015065
Merck Index:13,03851
Boiling Pt:269 °C (1013 hPa)
Melting Pt:–2 °C
Density:0.86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:138 °C
:228.38 g/mol
Storage Temperature:Ambient
EINECS:203-386-0
Frequently Bought Together