Order Entry
Austria
ContactUsLinkComponent
Ethyl laurate for synthesis, Sigma-Aldrich®
Ethyl laurate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ethyl laurate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Hill Formula:
    C₁₄H₂₈O₂
  • Chemical formula:
    CH₃(CH₂)₁₀COOC₂H₅
  • Molar Mass:
    228.37 g/mol
  • Boiling point:
    272 °C (1013 hPa)
  • Density:
    0.86 g/cm³ (20 °C)
  • Flash point:
    138 °C
  • Ignition temperature:
    300 °C
  • Melting Point:
    -2 °C
  • Vapor pressure:
    0.1 hPa (60 °C)
  • Refractive Index:
    1.4308 (20 °C, 589 nm)
  • Solubility:
    (20 °C) insoluble
  • LD 50 oral:
    LD50 Rat 5000 mg/kg
  • LD 50 dermal:
    LD50 Rabbit 5000 mg/kg
  • Storage class:
    10 Combustible liquids not in Storage Class 3
  • WGK:
    WGK 2 water endangering

Specification Test Results

Related Information

Frequently Bought Together