Supelco® Organic Reference Standard, Ethyl butyrate
Catalog # 75563-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:105-54-4
- Description:Ethyl butyrate
- Seq:02
- Storage conditions:2...30 °C
- Hazard class description:H226
- Shipping conditions:STD
- MDL No.:MFCD00009394
- M-Clarity quality level:MQ200
- Synonym(s):Butyric acid ethyl ester
- Melting point:–93 °C (lit.)
- Formula:CH₃CH₂CH₂C(O)OC₂H₅
- Molecular weight:116,16
- Density:0,875 g/ml at 25 °C (lit.)
- Flash point:26 °C
- Boiling point:120 °C (lit.)
Specifications
About this item
- GHS signal: GHS02
Caution: P210 - P233 - P240 - P241 - P242 - P243