
Paracetamol for synthesis, Sigma-Aldrich®
: Merck
:103-90-2
:
:C₈H₉NO₂
:MFCD00002328
:151.16 g/mol
:13,00048
:388 °C (1013 hPa)
:168 °C
:1.34 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:203-157-5
:GHS07
:Ambient
- Solubility:14 g/l (20 °C)
- Melting Point:169 - 171 °C
- Molar Mass:151.16 g/mol
- Boiling Point:500 °C
- Explosion Limit:15 %(V)
- Density:1.293 g/cm3 (21 °C)
- pH:5.5 - 6.5 (H2O, 20 °C) (saturated solution)
- Ignition Point:540 °C
Frequently Bought Together