Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetanilide

Acetanilide

Glass bottle for solids, 1 g
Artikelnr. PROOICRS0201
Irritant
Acetanilide
Artikelnr. PROOICRS0201
CAS Nummer:  103-84-4
Acetanilide

Glass bottle for solids, 1 g

CAS Nummer:  103-84-4
Preis
253,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • EINECS:203-150-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.