Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetanilide

Acetanilide

Glass bottle for solids, 500 mg
Catalog # PROOLGC2605
Irritant
Acetanilide
Catalog # PROOLGC2605
CAS Number:  103-84-4
Acetanilide

Glass bottle for solids, 500 mg

CAS Number:  103-84-4
Price
535,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • EINECS:203-150-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications