Order Entry
Austria
ContactUsLinkComponent
Adipohydrazide ≥97.0% for synthesis, Sigma-Aldrich®
Adipohydrazide ≥97.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Adipohydrazide ≥97.0% for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • CAS:
    1071-93-8
  • Hill Formula:
    C₆H₁₄N₄O₂
  • Molar Mass:
    174.2 g/mol
  • Flash point:
    less than 150 °C
  • Ignition temperature:
    360 °C
  • Melting Point:
    178 - 182 °C
  • Assay (HClO₄):
    ≥ 97,0 %(m)
  • Melting range (lower value):
    ≥ 178 °C
  • Melting range (upper value):
    ≤ 182 °C
  • Identity (IR):
    conforms

Specification Test Results

Related Information

Frequently Bought Together