Order Entry
Austria
Orders LinkContactUsLinkComponent
Urea ≥90%, TECHNICAL

Urea ≥90%, TECHNICAL

Cardboard carton, 25 kg
Catalog # 28875.460
Supplier:   VWR Chemicals
Urea ≥90%, TECHNICAL
Catalog # 28875.460
Supplier:  VWR Chemicals
CAS Number:  57-13-6
Urea ≥90%, TECHNICAL
Urea ≥90%, TECHNICAL

Cardboard carton, 25 kg

CAS Number:  57-13-6
Price
430,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specification Test Results

  • Assay:
    Min. 90 %

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.