Order Entry
Austria
ContactUsLinkComponent
Urea 8 mol/l in aqueous solution, Proteomics Grade
Urea 8 mol/l in aqueous solution, Proteomics Grade
Catalog # M124-250ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Urea 8 mol/l in aqueous solution, Proteomics Grade
Catalog # M124-250ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Clarity:
    Pass
  • Protease:
    none detected
  • Urea:
    7.8 – 8.2 Molar
  • Appearance:
    Clear and colorless liquid

Specification Test Results

About this item

 :
 :60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022