Order Entry
Austria
ContactUsLinkComponent
Formamide
Formamide
Catalog # ROTHP040.1
Supplier:  Roth Carl
CAS Number:  
undefined
Formamide
Catalog # ROTHP040.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Carbamaldehyde
 :
 :45.04 g/mol
Boiling Pt:210
EINECS:200-842-0
 :GHS08
Melting Pt:2
Storage Temperature:Refrigerator
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941
Density:1.1334 g/cm³ (20 °C)
Flash Pt:79
Merck Index:13,04261