
Formamide ≥99.5% (by total nitrogen basis) deionized for molecular biology
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:75-12-7
:
Formula:HCONH₂
MDL Number:MFCD00007941
Merck Index:13,04261
Boiling Pt:210 °C (1013 hPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
Flash Pt:79 °C
:45.04 g/mol
EINECS:200-842-0
:GHS08
Storage Temperature:Ambient
Specification Test Results
- DNases/RNases/Proteases:not detectable
- Assay (from N):min. 99.5 %
- Water:max. 0.1 %
- Chloride:max. 0.00005 %
- Fe:max. 0.00001 %
- PB:max. 0.00001 %
Specification Test Results
Frequently Bought Together