N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Catalog # 9222-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- Assay (HCON(CH₃)₂) (by GC, corrected for water):≥ 99.8 %
- Appearance:Passes Test
- Color (APHA):≤ 15
- Residue after Evaporation:≤ 0.0005 %
- Titrable Acid (meq/g):≤ 0.0005
- Titrable Base (meq/g):≤ 0.003
- Water (H₂O)(by Karl Fischer titrn):≤ 0.05 %
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Optical Absorbance (1-cm path vs water) - 270 nm:≤ 1.00
- Optical Absorbance (1-cm path vs water) - 275 nm:≤ 0.30
- Optical Absorbance (1-cm path vs water) - 295 nm:≤ 0.10
- Optical Absorbance (1-cm path vs water) - 310 nm:≤ 0.05
- Optical Absorbance (1-cm path vs water) - 340 nm:≤ 0.01
- Optical Absorbance (1-cm path vs water) - 400 nm:≤ 0.01
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:Management of Change (MOC) category = N
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284