Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates
undefined
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Residue on evaporation:
    Max. 0.0005 %
  • Water (K.F.):
    Max. 0.0050 %

Specification Test Results

Related Information

Frequently Bought Together