


N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- ACS - Assay (HCON(CH₃)₂) (by GC, corrected for water):≥99.8 %
- Appearance:Passes Test
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤0.005 %
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Titrable Base (meq/g):≤0.003
- ACS - Water (H₂O)(by Karl Fischer titrn):≤20 ppm
Specification Test Results
Frequently Bought Together