Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylformamide, BAKER ANALYZED® ACS, J.T.Baker®
N,N-Dimethylformamide, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
N,N-Dimethylformamide, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.8 %
  • Appearance:
    clear
  • Color (APHA):
    max. 15
  • Residue after Evaporation:
    max. 0.005 %
  • Titrable Acid (meq/g):
    max. 0.0005
  • Titrable Base (meq/g):
    max. 0.003
  • Water (H2O):
    max. 0.1 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together