


N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:14,3243
Boiling Pt:153 °C
Melting Pt:–61 °C
Density:0.944
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:57 °C (135 °F)
REACH:01-2119475605-32
:GHS08
:GHS02
:GHS07
Frequently Bought Together