Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylformamide 99.8% for peptide synthesis
N,N-Dimethylformamide 99.8% for peptide synthesis
Certificates
undefined
N,N-Dimethylformamide 99.8% for peptide synthesis

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (K.F.)
  • GC:
    ≥99.8 %
  • Color scale:
    ≤10 APHA
  • Free amine:
    ≤0.001 %
  • Acidity (CH3COOH):
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together