ContactUsLinkComponent
Electroporation system, square wave, ECM™ 830
Electroporation system, square wave, ECM™ 830
Supplier:  BTX
Certificates
undefined
Electroporation system, square wave, ECM™ 830
Supplier:  BTX
Specifications
  • Catalog No:
  • BTXI45-2052
  • BTXI45-0661
  • BTXI45-0662
  • Interfaces:
  • Touch screen interface
  • Touch screen interface
  • Touch screen interface
  • Charging time:
  • <7 s
  • <7 s
  • <7 s
  • Voltage range High Voltage Mode / 10 V resolution:
  • 505 – 3000 V
  • 505 - 3000 V
  • 505 - 3000 V
  • Voltage range Low Voltage Mode / 1 V resolution:
  • 5 - 500 V
  • 5 - 500V
  • 5 - 500V