Green 496 dUTP
Catalog # ENZOENZ42831
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 Nmo
- Description:Green 496 dUTP, 1 mM, 25 nmol
- Seq:02
Specifications
About this item
Green 496 [5-Fluorescein] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridisation procedures on fixed cells and tissues by direct fluorescence detection. Green 496 dUTP can also be used for multicolour fluorescence labeling.