Order Entry
Austria
ContactUsLinkComponent
Sodium pyruvate 0.1 mol/l in aqueous solution, sterile
Sodium pyruvate 0.1 mol/l in aqueous solution, sterile
Catalog # ICNA091682049
Supplier:  MP Biomedicals
CAS Number:  
undefined
Sodium pyruvate 0.1 mol/l in aqueous solution, sterile
Catalog # ICNA091682049
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :110.04 g/mol
Density:~1,267 g/ml (lit.)
Storage Temperature:Ambient
EINECS:204-024-4
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586