Order Entry
Austria
ContactUsLinkComponent
 
Flammable
Toxic
Di-tert-butyl pyrocarbonate
Catalog # MOLE62638603-100G
Supplier:  Molekula
CAS Number:  
undefined
Flammable
Toxic
Di-tert-butyl pyrocarbonate
Catalog # MOLE62638603-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :218.25 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:20…23 °C
Density:1.02 g/cm³ (20 °C)
Flash Pt:37 °C
Storage Temperature:Ambient
EINECS:246-240-1
UN:2930
ADR:6.1,I
 :GHS02
 :GHS06
CAS Number:24424-99-5
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805